Learn More
Fine Science Tools Artery Scissors, Ball Tip, Stainless Steel, 11.5 cm
SDP
Catalog No. NC9859722
Encompass
Tip Shape: Straight/ Tips: Sharp-Ball Tip/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No/ Feature: 1.5mm Ball Diameter
Catalog No. NC9859722 Supplier Fine Science Tools Supplier No. 1408011
Tip Shape: Straight/ Tips: Sharp-Ball Tip/ Cutting Edge: 25mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No/ Feature: 1.5mm Ball Diameter