Learn More
Fine Science Tools Vannas-Tübingen Spring Scissors, Straight, 8.5 cm
SDP
Catalog No. NC9280125
Encompass
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No
Catalog No. NC9280125 Supplier Fine Science Tools Supplier No. 1500308
Add to Cart
Add to Cart
Tip Shape: Straight/ Tips: Sharp/ Cutting Edge: 5mm/ Tip Diameter: 0.1mm/ Alloy / Material: Stainless Steel/ Length: 8.5cm/ Serrated: No