Learn More
Fine Science Tools SCISSORS
SDP
Catalog No. NC9389770
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 17cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9389770 Supplier Fine Science Tools Supplier No. 1411017
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 45mm/ Alloy / Material: Stainless Steel/ Length: 17cm/ Serrated: Yes/ Feature: ToughCut