Learn More
Roboz Surgical SCISSORS, 5", BLUNT / BLUNT, T
SDP
Catalog No. NC0564085
Encompass
Micro Dissecting Scissors; Straight; Blunt; Carbide Blades; 25mm Blade Length; 4-1/2 in. Overall Length
Catalog No. NC0564085 Supplier Roboz Surgical Supplier No. RS5984
Micro Dissecting Scissors; Straight; Blunt; Carbide Blades; 25mm Blade Length; 4-1/2 in. Overall Length