Learn More
Med Vet International Miltex Blepharoplasty Scissors, Tungsten Carbide, Curved, Serrated, German, 4.5in, Pm-6435
Catalog No. 502830766
Encompass_Preferred
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. Tungsten carbide guarantees durability. 4.5in in length. Curved blades. Made with high-end, medical grade German steel.
Catalog No. 50-283-0766 Supplier Med Vet International Supplier No. MILPM6435
Typically used to cut eye tissue for upper and lower blepharoplasty. The serrated blade ensures optimal grip of tissue, preventing slips. Tungsten carbide guarantees durability. 4.5in in length. Curved blades. Made with high-end, medical grade German steel.