Learn More
Fine Science Tools Bonn Strabismus Scissors - ToughCut, 9cm
SDP
Catalog No. NC9942722
Encompass
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: ToughCut
Catalog No. NC9942722 Supplier Fine Science Tools Supplier No. 1408509
Tip Shape: Curved/ Tips: Blunt-Blunt/ Cutting Edge: 14mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: Yes/ Feature: ToughCut