Learn More
Med Vet International Miltex Crown And Collar Scissors, Straight, 4.12in, V99D-133
Catalog No. 502829591
Encompass_Preferred
Used for restorative procedures such as cutting dental dam material, retraction cord and stainless steel crowns. Straight blades. 4.12in total length.
Catalog No. 50-282-9591 Supplier Med Vet International Supplier No. MILV99D133
Used for restorative procedures such as cutting dental dam material, retraction cord and stainless steel crowns. Straight blades. 4.12in total length.