Learn More
Med Vet International Miltex Crown And Collar Scissors, Curved, 4.12in, V99D-135
Catalog No. 502831953
Encompass_Preferred
Used for restorative procedures such as cutting dental dam material, retraction cord and stainless steel crowns. Curved blades. 4.12in total length.
Catalog No. 50-283-1953 Supplier Med Vet International Supplier No. MILV99D135
Used for restorative procedures such as cutting dental dam material, retraction cord and stainless steel crowns. Curved blades. 4.12in total length.