Learn More
Electron Microscopy Sciences Scissors with Curved Blades with Sharp/Blunt Points, 4½"
SDP
Catalog No. 5025556
Encompass_Preferred
Dissecting operating scissors. Made of stainless steel with one sharp point, one blunt point.
Points: Sharp/Blunt
Catalog No. 50-255-56 Supplier Electron Microscopy Sciences Supplier No. 7296402
Dissecting operating scissors. Made of stainless steel with one sharp point, one blunt point.
Points: Sharp/Blunt