Learn More
World Precision Instrument Dissecting Scissors, 12.5 cm, Curved
SDP
Catalog No. 50822345
Encompass
Ring-handled Dissecting Scissors are surgical scissors commonly used to cut and separate delicate tissues. These scissors are 10 or 12.5 cm long. These dissecting scissors are available with straight or curved blades. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel or premium German steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Catalog No. 50-822-345 Supplier World Precision Instrument Supplier No. 15923
Ring-handled Dissecting Scissors are surgical scissors commonly used to cut and separate delicate tissues. These scissors are 10 or 12.5 cm long. These dissecting scissors are available with straight or curved blades. Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries. Dissecting scissors have thinner (and often longer) blades than operating scissors. They are designed for cutting and probing in tight spaces. Their tips are pointed, where operating scissors are generally blunt. These are available in surgical grade stainless steel or premium German steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.