Learn More
Med Vet International Miltex Mayo-Noble Dissecting Scissors, Straight, Rounded Blades, German, 6.5in, 5-144
Catalog No. 503046478
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.5in in length. Curved rounded blades. Made with high-quality, medical grade German metal.
Catalog No. 50-304-6478 Supplier Med Vet International Supplier No. MIL5144
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.5in in length. Curved rounded blades. Made with high-quality, medical grade German metal.