Learn More
Med Vet International Miltex Mayo MeisterHand Dissecting Scissors, Straight, Standard Beveled Blades, German, 6.75in, MH5-124
Catalog No. 503046410
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Standard beveled straight blades. MeisterHand scissors guarantee superior quality. Made with high-quality, medical grade German metal.
Catalog No. 50-304-6410 Supplier Med Vet International Supplier No. MILMH5124
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Standard beveled straight blades. MeisterHand scissors guarantee superior quality. Made with high-quality, medical grade German metal.