Learn More
Med Vet International Miltex Medium Curve Enucleation Scissors, German, 5.12in, 18-1496
Catalog No. 502829144
Encompass_Preferred
Enucleation scissors are heavy duty scissors that are used for removing the globe from the eye. They are preferred by surgeons to dissect the connecting stalk as well as dissecting ocular muscles during ocular surgeries. Medium curve. 5.12in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-282-9144 Supplier Med Vet International Supplier No. MIL181496
Enucleation scissors are heavy duty scissors that are used for removing the globe from the eye. They are preferred by surgeons to dissect the connecting stalk as well as dissecting ocular muscles during ocular surgeries. Medium curve. 5.12in total length. Made with high-quality, medical-grade German steel.