Learn More
Med Vet International Miltex Full Curve Enucleation Scissors, German, 5in, 18-1498
Catalog No. 502643554
Encompass_Preferred
Enucleation scissors are heavy duty scissors that are used for removing the globe from the eye. They are preferred by surgeons to dissect the connecting stalk as well as dissecting ocular muscles during ocular surgeries. 5in in length. Made with high-quality, medical-grade German steel.
Catalog No. 50-264-3554 Supplier Med Vet International Supplier No. MIL181498
Enucleation scissors are heavy duty scissors that are used for removing the globe from the eye. They are preferred by surgeons to dissect the connecting stalk as well as dissecting ocular muscles during ocular surgeries. 5in in length. Made with high-quality, medical-grade German steel.