Learn More
Med Vet International Miltex Iris Scissors w/Probe Points, Angled On Side, German, 4.5in, 5-310
Catalog No. 502832259
Encompass_Preferred
Operating room grade ultra-fine sharp-tip scissors. Ideal for ophthalmic, corneal, and optical applications. Made with the highest quality German stainless steel. Probe points allow for easy examination and maneuverability. 4.5in total length. Designed for intraocular operations. Made with German steel.
Catalog No. 50-283-2259 Supplier Med Vet International Supplier No. MIL5310
Operating room grade ultra-fine sharp-tip scissors. Ideal for ophthalmic, corneal, and optical applications. Made with the highest quality German stainless steel. Probe points allow for easy examination and maneuverability. 4.5in total length. Designed for intraocular operations. Made with German steel.