Learn More
Med Vet International Miltex Kilner Scissors, Straight Fine Points, German, 4.5in, PM-0448
Catalog No. 503046763
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Ultra sharp SuperCut blades for ease of use with the durability of tungsten carbide. Straight blades with fine points. 4.5in total length. Made with high-quality, high-end German steel.
Catalog No. 50-304-6763 Supplier Med Vet International Supplier No. MILPM0448
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Ultra sharp SuperCut blades for ease of use with the durability of tungsten carbide. Straight blades with fine points. 4.5in total length. Made with high-quality, high-end German steel.