Learn More
Med Vet International Miltex Foster Needle Holder & Scissors, Straight Smooth, German, 4.25in, Pm-0438
Catalog No. 502830480
Encompass_Preferred
A needle holder, also called needle driver, is a surgical instrument, similar to a hemostat, used by doctors and surgeons to hold a suturing needle for closing wounds during suturing and surgical procedures. Straight blades with a smoothness to them. 4.25in in length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0480 Supplier Med Vet International Supplier No. MILPM0438
A needle holder, also called needle driver, is a surgical instrument, similar to a hemostat, used by doctors and surgeons to hold a suturing needle for closing wounds during suturing and surgical procedures. Straight blades with a smoothness to them. 4.25in in length. Made with high-quality, medical-grade German steel.