Learn More
Med Vet International Miltex Gorney Rhytidectomy Scissors, Curved, Supercut, German, 7.25in, Pm-6831
Catalog No. 502830088
Encompass_Preferred
Rhytidectomy scissors offer extended shaft lengths for greater surgical visualization. Featured are specially-designed razor sharp upper blade edges to cut cleanly and quickly though dense or delicate tissue. Curved SuperCut blades. 7.25in in length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-0088 Supplier Med Vet International Supplier No. MILPM6831
Rhytidectomy scissors offer extended shaft lengths for greater surgical visualization. Featured are specially-designed razor sharp upper blade edges to cut cleanly and quickly though dense or delicate tissue. Curved SuperCut blades. 7.25in in length. Made with high-quality, medical grade German steel.