Learn More
Med Vet International Miltex Supercut Gradle Scissors, 3.75in, Curved
Supplier:  Med Vet International MIL18SC1652
Encompass_Preferred
The Miltex SuperCut 18-SC-1652 operating scissors are made of German grade stainless steel and have a length of 3.75in. These operating scissors feature a black handle with a razor sharp edge and 2 sharp tips.
Catalog No. 50-264-3552