Learn More
Med Vet International Miltex Gradle Eye Suture Scissors, Slightly Curved, Sharp Points, 3.75in, 18-1652
Supplier:  Med Vet International MIL181652
Encompass_Preferred
Ophthalmic and eye scissors feature small blades, a traditional ring-handle style. Use for cutting into and probing delicate optical tissue. Slightly curved blades with sharp points. 3.75in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-282-9147