Learn More
World Precision Instrument Iris Scissors, 11.5 cm, Curved
SDP
Catalog No. 50822801
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-801 Supplier World Precision Instrument Supplier No. 501759
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.