Learn More
Med Vet International Miltex Iris Scissors, 3.5in., Curved
Catalog No. 502830068
Encompass_Preferred
The Miltex 5-302 Iris Scissors have a length of 3.5in. Iris Scissors are a type of surgical scissor with short blades and is commonly used for ophthalmic surgery. This 3.5in. curved Iris Scissors are made of German grade stainless steel.
Catalog No. 50-283-0068 Supplier Med Vet International Supplier No. MIL5302
The Miltex 5-302 Iris Scissors have a length of 3.5in. Iris Scissors are a type of surgical scissor with short blades and is commonly used for ophthalmic surgery. This 3.5in. curved Iris Scissors are made of German grade stainless steel.