Learn More
Med Vet International Miltex Iris Scissors, 4.5in., Curved, Tungsten Carbide
Catalog No. 502831325
Encompass_Preferred
The Miltex 5-306TC Tungsten Carbide Iris Scissors have a length of 4.5in. Iris Scissors are a type of surgical scissor with short blades and is commonly used for ophthalmic surgery. This 4.5in. curved Iris Scissors are made of German grade stainless steel and Tungsten Carbide to enhance their performance and longevity.
Catalog No. 50-283-1325 Supplier Med Vet International Supplier No. MIL5306TC
The Miltex 5-306TC Tungsten Carbide Iris Scissors have a length of 4.5in. Iris Scissors are a type of surgical scissor with short blades and is commonly used for ophthalmic surgery. This 4.5in. curved Iris Scissors are made of German grade stainless steel and Tungsten Carbide to enhance their performance and longevity.