Learn More
Med Vet International 4.5-in Economy Iris Scissors, Curved
Catalog No. 502224066
Encompass_Preferred
Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.
Catalog No. 50-222-4066 Supplier Med Vet International Supplier No. 4331
Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.