Learn More
Fine Science Tools Fine Scissors, Stainless Steel, Sharp, 9 cm
SDP
Catalog No. NC9942723
Encompass
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No
Catalog No. NC9942723 Supplier Fine Science Tools Supplier No. 1406309
Tip Shape: Angled to Side/ Tips: Sharp-Sharp/ Cutting Edge: 22mm/ Alloy / Material: Stainless Steel/ Length: 9cm/ Serrated: No