Learn More
Med Vet International Miltex Meisterhand Iris Scissors, Curved, Supercut, German, 4.5in, Mh5-Sc-306
Catalog No. 502832253
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. SuperCut blades allow for easy use. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. MeisterHand scissors guarantee exceptional craftsmanship. Curved SuperCut blades ensure clean cuts. 4.5in total length. Made with high-quality, high-end German steel.
Catalog No. 50-283-2253 Supplier Med Vet International Supplier No. MILMH5SC306
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. SuperCut blades allow for easy use. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. MeisterHand scissors guarantee exceptional craftsmanship. Curved SuperCut blades ensure clean cuts. 4.5in total length. Made with high-quality, high-end German steel.