Learn More
Med Vet International Miltex Iris Scissors, Curved, Tungsten Carbide, Sharp/Sharp, German, 4.5in, Pm-2705
Supplier: Med Vet International MILPM2705
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. SuperCut blades allow for easy use. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Tungsten coating keeps the instrument durable. Curved sharp/sharp pointed blades. 4.5in total length. Made with high-quality, high-end German steel.