Learn More
Med Vet International Miltex MeisterHand Iris Scissors, Curved, Standard Flat Shanks, German, 4in, MH18-1416
Catalog No. 503046777
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Fabricated with MeisterHand signature craftsmanship. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. 4in total. Made with high-quality, high-end German steel.
Catalog No. 50-304-6777 Supplier Med Vet International Supplier No. MILMH181416
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Fabricated with MeisterHand signature craftsmanship. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. 4in total. Made with high-quality, high-end German steel.