Learn More
World Precision Instrument Ribbon Handle Iris Scissors, 11cm, Curved, Large Ring
SDP
Supplier:  World Precision Instrument 501768
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-810