Learn More
Med Vet International Miltex Meisterhand Iris Scissors, Straight, 20mm Blades, Delicate, German, 3.5in, Mh18-1396
Supplier: Med Vet International MILMH181396
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. 3.5in in length. MeisterHand provide craftsmanship that's hard to match. Straight blades with a delicate finish. 20mm blades. 3.5in total length. Made with high-quality, high-end German steel.