Learn More
Med Vet International Miltex Iris Scissors, Straight, Fine, Sharp Points, German, 4in, MH5-300
Catalog No. 502643486
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. MeisterHand scissors provides signature exceptional craftsmanship. Straight fine blades with sharp points. 4in total length. Made with high-quality, high-end German steel.
Catalog No. 50-264-3486 Supplier Med Vet International Supplier No. MILMH5300
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. MeisterHand scissors provides signature exceptional craftsmanship. Straight fine blades with sharp points. 4in total length. Made with high-quality, high-end German steel.