Learn More
Med Vet International Miltex Iris Scissors, Straight, 4.5in, 12 Pack, EG5-304
Catalog No. 502643500
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight blades. 4.5in total length. Made with high-quality, high-end German steel.
Catalog No. 50-264-3500 Supplier Med Vet International Supplier No. MILEG5304
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. Applicable in a multitude of environments such as, dental, ophthalmic, oral, and more. Straight blades. 4.5in total length. Made with high-quality, high-end German steel.