Learn More
World Precision Instrument Iris Scissors, 12.5cm, SuperCut, Straight
SDP
Catalog No. 50822300
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-300 Supplier World Precision Instrument Supplier No. 14225
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.