Learn More
World Precision Instrument Iris Scissors, 11cm, Curved, Tungsten Carbide, German
SDP
Catalog No. 50822382
Encompass
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.
Catalog No. 50-822-382 Supplier World Precision Instrument Supplier No. 500217G
Iris scissors were originally designed for ophthalmic procedures but are now used in a wide range of applications. The tips are sharp, and they come in curved, straight or angled styles.