Learn More
Med Vet International Miltex MeisterHand Iris Scissors, Tungsten Carbide, Straight, Fine, Sharp Points, German, 4.62in, MH5-304TC
Catalog No. 502643485
Encompass_Preferred
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. MeisterHand scissors guarantee exceptional craftsmanship. Ultra sharp SuperCut blades for ease of use with the durability of tungsten carbide. Straight fine blades with sharp points. 4.62in total length. Made with high-quality, high-end German steel.
Catalog No. 50-264-3485 Supplier Med Vet International Supplier No. MILMH5304TC
Universal scissors for cutting sutures, tissue, gauze, bandages, and more. MeisterHand scissors guarantee exceptional craftsmanship. Ultra sharp SuperCut blades for ease of use with the durability of tungsten carbide. Straight fine blades with sharp points. 4.62in total length. Made with high-quality, high-end German steel.