Learn More
Med Vet International Miltex Kelly Scissors, Curved, Sharp Points, German, 6.25in, 5-252
Catalog No. 502830955
Encompass_Preferred
Used to quickly and efficiently cut and excise the excess fascia or dead tissue. Curved blades with sharp points. 6.25in total length. Made with high-quality, medical grade German steel.
Catalog No. 50-283-0955 Supplier Med Vet International Supplier No. MIL5252
Used to quickly and efficiently cut and excise the excess fascia or dead tissue. Curved blades with sharp points. 6.25in total length. Made with high-quality, medical grade German steel.