Learn More
Med Vet International Miltex Knowles Bandage Scissors, Angled, German, 5.5in, Mh5-561
Catalog No. 502831353
Encompass_Preferred
Durable, lightweight, dependable, and designed for cutting thick dressing material like bandages or gauze. Angled blades to allow access to hard to reach places. 5.5in in length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-1353 Supplier Med Vet International Supplier No. MILMH5561
Durable, lightweight, dependable, and designed for cutting thick dressing material like bandages or gauze. Angled blades to allow access to hard to reach places. 5.5in in length. Made with high-quality, medical-grade German steel.