Learn More
Fine Science Tools SCISSORS LG-LOOPED W/BALL FINE
SDP
Catalog No. NC9216476
Encompass
Tip Shape: Angled to Side/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.125mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No
Catalog No. NC9216476 Supplier Fine Science Tools Supplier No. 1500609
Tip Shape: Angled to Side/ Tips: Sharp/ Cutting Edge: 10mm/ Tip Diameter: 0.125mm/ Alloy / Material: Stainless Steel/ Length: 10cm/ Serrated: No