Learn More
World Precision Instrument Spencer Stitch Scissors, 10.75cm, German
SDP
Supplier:  World Precision Instrument 501703G
Encompass
Spencer Stitch Scissors are stitch-cutting scissors or suture scissors for removing sutures or stitches. Spencer Stitch Scissors have a relatively short, straight, and narrow blades. The tips of Spencer stitch scissors are blunt rather than sharp to minimize the risk of accidentally puncturing or damaging underlying tissues while cutting sutures. One blade is notched to provide improved grip and traction when cutting through sutures, preventing slippage and ensuring clean cuts of just the suture. The ring handles give you a firm, comfortable grip for precision control. They come in a variety of lengths, from 9.0 cm to 13.25 cm. Shorter handles typically offer more control, and longer handles give you greater reach. These Spencer Stitch Scissors are manufactured from surgical grade stainless steel or premium German steel, both of which are autoclavable. WPI recommends properly cleaning all your surgical instruments promptly after use.
Catalog No. 50-822-708