Learn More
Med Vet International Miltex Littauer Scissors, Standard, German, 5.5in, Mh9-104
Catalog No. 502830335
Encompass_Preferred
Littauer scissors feature a hook-shaped blade that easily slides under a suture without it slipping off the end of the blade as it is being cut. These suture scissors are commonly used in both human medicine and veterinary. medicine, as they serve the same purpose in both situations. 5.5in total length. Standard issue. Made with high-quality, medical-grade German steel
Catalog No. 50-283-0335 Supplier Med Vet International Supplier No. MILMH9104
Littauer scissors feature a hook-shaped blade that easily slides under a suture without it slipping off the end of the blade as it is being cut. These suture scissors are commonly used in both human medicine and veterinary. medicine, as they serve the same purpose in both situations. 5.5in total length. Standard issue. Made with high-quality, medical-grade German steel