Learn More
Med Vet International Miltex Littauer Jr. Scissors, Light Pattern, German, 4.75in, Mh9-102
Catalog No. 502830358
Encompass_Preferred
Littauer scissors feature a hook-shaped blade that easily slides under a suture without it slipping off the end of the blade as it is being cut. These suture scissors are commonly used in both human medicine and veterinary. medicine, as they serve the same purpose in both situations. 4.75in total length. Light pattern. Made with high-quality, medical-grade German steel
Catalog No. 50-283-0358 Supplier Med Vet International Supplier No. MILMH9102
Littauer scissors feature a hook-shaped blade that easily slides under a suture without it slipping off the end of the blade as it is being cut. These suture scissors are commonly used in both human medicine and veterinary. medicine, as they serve the same purpose in both situations. 4.75in total length. Light pattern. Made with high-quality, medical-grade German steel