Learn More
Med Vet International Miltex Martin Cartilage Scissors, Curved, Serrated Non-Slip Cutting Edge, German, 5.62in, Mh27-1000
Catalog No. 502831904
Encompass_Preferred
Scissors used in rhinoplasty procedures. The small, curved nature of the blade allows the surgeon to effectively separate the alar cartilage from the superficial facia to either trim or modify its contour. 5.62in length. Curved blades with non-slip serrated edges. Made out of high-quality, medical-grade steel.
Catalog No. 50-283-1904 Supplier Med Vet International Supplier No. MILMH271000
Scissors used in rhinoplasty procedures. The small, curved nature of the blade allows the surgeon to effectively separate the alar cartilage from the superficial facia to either trim or modify its contour. 5.62in length. Curved blades with non-slip serrated edges. Made out of high-quality, medical-grade steel.