Learn More
Med Vet International Miltex Dissecting Scissors, 6.75in., Straight
Catalog No. 502831187
Encompass_Preferred
The Miltex 5-124 Mayo dissecting scissors are made of German grade stainless steel. They have a length of 6.75in., a straight bevel, and 2 blunt tips.
Catalog No. 50-283-1187 Supplier Med Vet International Supplier No. MIL5124
The Miltex 5-124 Mayo dissecting scissors are made of German grade stainless steel. They have a length of 6.75in., a straight bevel, and 2 blunt tips.