Learn More
Med Vet International Miltex Mayo Scissors, Supercut, Curved, One Serrated Blade, German, 6.75in, 5-Sc-126
Catalog No. 502832243
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Curved SuperCut blades with serrated edges to make dissection and biopsies easy. Made with high-quality, medical grade German metal.
Catalog No. 50-283-2243 Supplier Med Vet International Supplier No. MIL5SC126
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Curved SuperCut blades with serrated edges to make dissection and biopsies easy. Made with high-quality, medical grade German metal.