Learn More
Med Vet International Miltex Mayo Dissecting Scissors, Curved, Tungsten, Standard Beveled Blades, German, 5.75in, 5-122Tc
Catalog No. 502831186
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.75in in length. Standard beveled curved blades. Tungsten carbide coated for increased durability. Made with high-quality, medical grade German metal.
Catalog No. 50-283-1186 Supplier Med Vet International Supplier No. MIL5122TC
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.75in in length. Standard beveled curved blades. Tungsten carbide coated for increased durability. Made with high-quality, medical grade German metal.