Learn More
Med Vet International Miltex Mayo Scissors, Straight, Blunt, German, 5.62in, Pm-0460
Catalog No. 502830481
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.62in in length. Straight blades with blunt points making it easier for dissection and biopsy. Made with high-quality, medical grade German metal.
Catalog No. 50-283-0481 Supplier Med Vet International Supplier No. MILPM0460
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 5.62in in length. Straight blades with blunt points making it easier for dissection and biopsy. Made with high-quality, medical grade German metal.