Learn More
Med Vet International Miltex Mayo Scissors, Supercut, Straight, One Serrated Blade, German, 6.75in, 5-Sc-124
Catalog No. 502832242
Encompass_Preferred
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Straight SuperCut blades with a serrated edge to make dissections and biopsies easy. Made with high-quality, medical grade German metal.
Catalog No. 50-283-2242 Supplier Med Vet International Supplier No. MIL5SC124
Thicker and more heavy duty than most, Mayo scissors are generally used near the surface of a surgical wound to cut fascia and muscle layers. It is standard for Mayo scissors to have standard or extra-long shafts. 6.75in in length. Straight SuperCut blades with a serrated edge to make dissections and biopsies easy. Made with high-quality, medical grade German metal.