Learn More
World Precision Instrument Mayo Scissors, 14 cm, Curved, Tungsten Carbide, German
SDP
Supplier:  World Precision Instrument 500219G
Encompass
Tungsten Carbide Mayo Scissors, also called Mayo Dissecting Scissors, are surgical scissors made popular by the Mayo Clinic. They have ring handles and long, straight, thick blades with semi-blunt tips that are straight or curved. The surgical scissors vary in length from 14 cm (5.5") to 17.5 cm (7"). Longer blades give you greater reach for deeper dissection. The blades are sturdy for heavy duty work like cutting connective tissues, fascia, and sutures. Mayo Scissors are often used for dissection, where the blunt tips prevent accidental damage to surrounding tissue and may be used to delicately probe. They may also be used for dressing and bandage removal. 
Catalog No. 50-822-386