Learn More
World Precision Instrument McPherson-Vannas Spring Scissors, 8cm, Curved, 5mm Blades, German
SDP
Catalog No. 50822579
Encompass
The McPherson-Vannas spring scissors were originally designed by Dr. David McPherson and Dr. Erkki Vannas in the 1970s specifically for microsurgery, which requires precision in the dissection of delicate tissues. The fine, sharp tips of these Vannas spring scissors allow for precise cutting and dissection of small tissues. For greater visibility at the tip, select the curved blades. These surgical scissors have a smooth cutting action for a clean cut with minimal damage to surrounding tissues. The lightweight and ergonomic design of the McPherson-Vannas spring scissors reduces hand fatigue during use. Like all our spring scissors, these Vannas spring scissors are ideal for ambidextrous use and are often preferred by left-handed operators. These scissors are crafted of surgical grade stainless steel or our premium German steel, ensuring their durability. These surgical scissors can be sterilized with a chemical sterilant or by autoclaving.
Catalog No. 50-822-579 Supplier World Precision Instrument Supplier No. 501234G
The McPherson-Vannas spring scissors were originally designed by Dr. David McPherson and Dr. Erkki Vannas in the 1970s specifically for microsurgery, which requires precision in the dissection of delicate tissues. The fine, sharp tips of these Vannas spring scissors allow for precise cutting and dissection of small tissues. For greater visibility at the tip, select the curved blades. These surgical scissors have a smooth cutting action for a clean cut with minimal damage to surrounding tissues. The lightweight and ergonomic design of the McPherson-Vannas spring scissors reduces hand fatigue during use. Like all our spring scissors, these Vannas spring scissors are ideal for ambidextrous use and are often preferred by left-handed operators. These scissors are crafted of surgical grade stainless steel or our premium German steel, ensuring their durability. These surgical scissors can be sterilized with a chemical sterilant or by autoclaving.