Learn More
World Precision Instrument Metzenbaum Scissors, 14.5 cm, Curved, German
SDP
Catalog No. 50822611
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • Stainless steel
Catalog No. 50-822-611 Supplier World Precision Instrument Supplier No. 501253G
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
  • Stainless steel